missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polydisperse Particle Standard Certified Reference Material, Particle Size Standard, PS314, 10-100 μm, MilliporeSigma™ Supelco™
Specifications
Specifications
| Physical Form | Particles |
| Quantity | 10 x 0.5 g |
| Grade | Certified Reference Material |