missing translation for 'onlineSavingsMsg'
Learn More

Polycaprolactone, M.W. approx. 80,000

Catalog No. p-7214821
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC178301000 100 g
AC178305000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC178301000 Supplier Thermo Scientific Chemicals Supplier No. 178301000
Only null left

CAS: 24980-41-4 | (C6H10O2)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24980-41-4
Molecular Formula (C6H10O2)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00084404
InChI Key XNCNNDVCAUWAIT-UHFFFAOYSA-N
IUPAC Name oxepan-2-one
SMILES *-CCCCCC(=O)O-*

Specifications

Melting Point 60.0°C
Linear Formula [-O(CH2)5CO-]n
Quantity 100 g
Chemical Name or Material Polycaprolactone

RUO – Research Use Only