accessibility menu, dialog, popup

UserName

∗O∗On

CAS RN 24980-41-4

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C6H10O2)n
InChi Key: —
SMILES: *-CCCCCC(=O)O-*
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results