missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Polyanetholesulfonic acid sodium salt
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
$251.31 - $251.31
Specifications
| CAS | 55963-78-5 |
|---|---|
| MDL Number | MFCD00148427 |
| Synonym | Sodium polyanetholesulfonate |
| Molecular Weight (g/mol) | 9,000-11,000 |
| Biological Activity | Respiratory system |