missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Polyanetholesulfonic acid sodium salt

Catalog No. 44446425G
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44446425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44446425G Supplier Sigma Aldrich Supplier No. 44446425G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Anticoagulant

Specifications

CAS 55963-78-5
Quantity 25 g
MDL Number MFCD00148427
Synonym Sodium polyanetholesulfonate
Molecular Weight (g/mol) 9,000-11,000
Biological Activity Respiratory system