missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 20000-30000
CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol
Supplier: Thermo Scientific Chemicals 396760025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
| White to Yellow | |
| 4.5 to 6.5 (4% aq. soln.) | |
| 9002-89-5 | |
| 90%% (Na2O) max. | |
| [-CH2CH(OH)-]n | |
| vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval | |
| 2.5 kg | |
| OC(-*)C-* | |
| 44.05 | |
| 87.0 to 89.0% (degree of hydrolysis) | |
| Plastic Bottle |
| Poly(vinyl alcohol) | |
| Powder | |
| 160.0°C to 240.0°C | |
| (C2H4O)n | |
| MFCD00081922 | |
| Solubility in water: soluble in hot water. | |
| IMROMDMJAWUWLK-UHFFFAOYSA-N | |
| ethenol | |
| 11199 | |
| Authentic | |
| 3.5 to 4.5cP (4% aq. solution, 20°C) Brookfield |
Chemical Identifiers
| 9002-89-5 | |
| 44.05 | |
| IMROMDMJAWUWLK-UHFFFAOYSA-N | |
| 11199 | |
| OC(-*)C-* |
| (C2H4O)n | |
| MFCD00081922 | |
| vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval | |
| ethenol |
RUO – Research Use Only