missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 20000-30000

Catalog No. AC396760250
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC396760250 25 g Plastic bottle
AC396765000 500 g Plastic bottle
AC396760010 1 kg Plastic bottle
AC396760025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC396760250 Supplier Thermo Scientific Chemicals Supplier No. 396760250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
PubChem CID 11199
IUPAC Name ethenol
SMILES OC(-*)C-*

Specifications

Color White to Yellow
pH 4.5 to 6.5 (4% aq. soln.)
Physical Form Powder
Melting Point 160.0°C to 240.0°C
Ash 90%% (Na2O) max.
Quantity 25 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Linear Formula [-CH2CH(OH)-]n
Viscosity 3.5 to 4.5cP (4% aq. solution, 20°C) Brookfield
Solubility Information Solubility in water: soluble in hot water.
Percent Purity 87.0 to 89.0% (degree of hydrolysis)
Chemical Name or Material Poly(vinyl alcohol)
Show More Show Less

RUO – Research Use Only