missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl Alcohol) (n=approx. 2,000) (degree of saponification ca. 80mol%), TCI America™

Catalog No. p080425g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P080425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P080425G Supplier TCI America Supplier No. P080425G
Only null left

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
PubChem CID 11199
IUPAC Name ethenol
SMILES OC(-*)C-*

Specifications

Color White-Yellow
Physical Form Crystal-Powder at 20°C
Quantity 25 g
Chemical Name or Material Poly(vinyl Alcohol) (n=approx. 2,000) (degree of saponification ca. 80mol%)
EINECSNumber (6)-0682
RTECSNumber TR8100000
TSCA Yes