missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(vinyl Alcohol) n=1750+/-50, TCI America™
$86.64 - $173.26
Chemical Identifiers
| CAS | 9002-89-5 |
|---|---|
| Molecular Formula | (C2H4O)n |
| Molecular Weight (g/mol) | 44.05 |
| MDL Number | MFCD00081922 |
| InChI Key | IMROMDMJAWUWLK-UHFFFAOYSA-N |
| Synonym | vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval |
| PubChem CID | 11199 |
| IUPAC Name | ethenol |
| SMILES | OC(-*)C-* |
Chemical Identifiers
| 9002-89-5 | |
| 44.05 | |
| IMROMDMJAWUWLK-UHFFFAOYSA-N | |
| 11199 | |
| OC(-*)C-* |
| (C2H4O)n | |
| MFCD00081922 | |
| vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval | |
| ethenol |
Specifications
| White | |
| 9002-89-5 | |
| MFCD00081922 | |
| 25 g | |
| OC(-*)C-* | |
| 44.05 | |
| Poly(vinyl Alcohol) n=1750+/-50 |
| Crystal-Powder at 20°C | |
| (C2H4O)n | |
| vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval | |
| IMROMDMJAWUWLK-UHFFFAOYSA-N | |
| ethenol | |
| 11199 |
Safety and Handling
EINECSNumber : (6)-0682
RTECSNumber : TR8100000
TSCA : Yes