missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl Alcohol) n=1750+/-50, TCI America™

Catalog No. P0469500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0469500G 500 g
P046925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0469500G Supplier TCI America Supplier No. P0469500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
PubChem CID 11199
IUPAC Name ethenol
SMILES OC(-*)C-*

Specifications

Color White
Physical Form Crystal-Powder at 20°C
Quantity 500 g
Chemical Name or Material Poly(vinyl Alcohol) n=1750+/-50
EINECSNumber (6)-0682
RTECSNumber TR8100000
TSCA Yes