missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 85,000 to 120,000, Thermo Scientific Chemicals

Catalog No. p-4813608
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC450145000 500 g Plastic bottle
AC450140010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC450145000 Supplier Thermo Scientific Chemicals Supplier No. 450145000
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
PubChem CID 11199
IUPAC Name ethenol
SMILES OC(-*)C-*

Specifications

Melting Point 230.0°C to 240.0°C
Solubility Information Solubility in water: soluble in hot water.
Quantity 500 g
Packaging Plastic bottle
Chemical Name or Material Poly(vinyl alcohol)

RUO – Research Use Only