missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 31,000-50,000
Indicator | CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol
$44.51 - $44.51
Chemical Identifiers
| CAS | 9002-89-5 |
|---|---|
| Molecular Formula | (C2H4O)n |
| Molecular Weight (g/mol) | 44.05 |
| MDL Number | MFCD00081922 |
| InChI Key | IMROMDMJAWUWLK-UHFFFAOYSA-N |
| Synonym | vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval |
| PubChem CID | 11199 |
| IUPAC Name | ethenol |
| SMILES | OC(-*)C-* |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC183141000
|
Thermo Scientific Chemicals
183141000 |
100 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 9002-89-5 | |
| 44.05 | |
| IMROMDMJAWUWLK-UHFFFAOYSA-N | |
| 11199 | |
| OC(-*)C-* |
| (C2H4O)n | |
| MFCD00081922 | |
| vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval | |
| ethenol |
Specifications
| 9002-89-5 | |
| 4.5 to 6.5 (4% aq. soln.) | |
| (C2H4O)n | |
| MFCD00081922 | |
| 15,7706 | |
| Solubility in water: soluble in hot water. | |
| OC(-*)C-* | |
| 44.05 | |
| 5.2-6.2 mPa.s (20°C 4% aq. sol.) | |
| Poly(vinyl alcohol) |
| 230.0°C to 240.0°C | |
| Authentic | |
| [-CH2CH(OH)-]n | |
| 100 g | |
| vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval | |
| IMROMDMJAWUWLK-UHFFFAOYSA-N | |
| ethenol | |
| 11199 | |
| 87.0-89% |
RUO – Research Use Only