missing translation for 'onlineSavingsMsg'
Learn More

Poly(styrenesulfonic acid sodium salt) for for GPC 4,300, MilliporeSigma™ Supelco™

Catalog No. 111015880
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015880 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015880 Supplier Merck Emd Millipore Supplier No. 81606250MG

Specifications

CAS 9080-79-9
Physical Form Neat
MDL Number MFCD00084449
Synonym Poly(sodium styrenesulfonate)
Type Analytical Standard
Quantity 250 mg
Grade GPC