missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(styrenesulfonic acid sodium salt) for for GPC 17,000, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 9080-79-9 |
| Physical Form | Neat |
| MDL Number | MFCD00084449 |
| Synonym | Poly(sodium styrenesulfonate) |
| Type | Analytical Standard |
| Quantity | 250 mg |
| Grade | GPC |