missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(N-vinylcarbazole), Secondary Standard, W.W. 135,6000, M.N. 56,400
CAS: 25067-59-8 | (C14H11N)n | 193.25 g/mol
$179.36 - $179.36
Chemical Identifiers
| CAS | 25067-59-8 |
|---|---|
| Molecular Formula | (C14H11N)n |
| Molecular Weight (g/mol) | 193.25 |
| MDL Number | MFCD00134336 |
| InChI Key | KKFHAJHLJHVUDM-UHFFFAOYSA-N |
| Synonym | 9-vinylcarbazole, n-vinylcarbazole, 9-vinyl-9h-carbazole, 9h-carbazole, 9-ethenyl, n-vinyl carbazole, vinylcarbazole, 1-vinylcarbazole, poly 9-vinylcarbazole, carbazole, 9-vinyl, poly-n-vinylcarbazole |
| PubChem CID | 15143 |
| SMILES | *-CC(-*)N1C2=C(C=CC=C2)C2=C1C=CC=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC329690050
|
Thermo Scientific Chemicals
329690050 |
5 g |
Each for $179.36
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 25067-59-8 | |
| 193.25 | |
| KKFHAJHLJHVUDM-UHFFFAOYSA-N | |
| 15143 |
| (C14H11N)n | |
| MFCD00134336 | |
| 9-vinylcarbazole, n-vinylcarbazole, 9-vinyl-9h-carbazole, 9h-carbazole, 9-ethenyl, n-vinyl carbazole, vinylcarbazole, 1-vinylcarbazole, poly 9-vinylcarbazole, carbazole, 9-vinyl, poly-n-vinylcarbazole | |
| *-CC(-*)N1C2=C(C=CC=C2)C2=C1C=CC=C2 |
Specifications
| White | |
| 25067-59-8 | |
| MFCD00134336 | |
| 9-vinylcarbazole, n-vinylcarbazole, 9-vinyl-9h-carbazole, 9h-carbazole, 9-ethenyl, n-vinyl carbazole, vinylcarbazole, 1-vinylcarbazole, poly 9-vinylcarbazole, carbazole, 9-vinyl, poly-n-vinylcarbazole | |
| 5 g | |
| *-CC(-*)N1C2=C(C=CC=C2)C2=C1C=CC=C2 | |
| 193.25 | |
| Authentic | |
| Glass bottle | |
| 1.2 |
| Crystalline Powder | |
| (C14H11N)n | |
| 1.2000g/mL | |
| Solubility in water: insoluble. Other solubilities: soluble in benzene, chloroform, dichloromethane,, dioxane, thf, toluene | |
| KKFHAJHLJHVUDM-UHFFFAOYSA-N | |
| 9-ethenyl-9H-carbazole | |
| 15143 | |
| 3% max. | |
| 1.683 | |
| Poly(N-vinylcarbazole), Secondary standard |
Safety and Handling
RTECSNumber : FE6225480
TSCA : TSCA
RUO – Research Use Only