missing translation for 'onlineSavingsMsg'
Learn More

Poly(methyl Vinyl Ether) (30% in Water), TCI America™

Catalog No. P0384500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0384500G 500 g
P038425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0384500G Supplier TCI America Supplier No. P0384500G
Only null left

Chemical Identifiers

CAS 9003-09-2
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00161528
InChI Key XJRBAMWJDBPFIM-UHFFFAOYSA-N
Synonym Methyl Vinyl Ether Polymer, Poly(vinyl Methyl Ether), Vinyl Methyl Ether Polymer
PubChem CID 7861
IUPAC Name methoxyethene
SMILES COC=C

Specifications

Quantity 500 g
Physical Form Liquid
Chemical Name or Material Poly(methyl Vinyl Ether) (30% in Water)
EINECSNumber (6)-0982
TSCA Yes