missing translation for 'onlineSavingsMsg'
Learn More

Poly(methyl methacrylate) for for GPC 10,000, MilliporeSigma™ Supelco™

Catalog No. 111015839
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015839 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015839 Supplier Merck Emd Millipore Supplier No. 81497500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 9011-14-7
Ignition Point 580°F
Density 1.200 g/cm3
Flash Point >250°C
Physical Form Neat
Linear Formula [CH2 C(CH3)(CO2 CH3)]n
MDL Number MFCD00134349
Synonym PMMA; Poly(methacrylic acid methyl ester)
Solubility Insoluble in water
Type Analytical Standard
Quantity 500 mg
Grade GPC
Show More Show Less