missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Poly-L-Lysine Solution

Catalog No. a005c
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A005C 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A005C Supplier MilliporeSigma™ Supplier No. A005C
Only null left

Used as coating to enhance cell attachment and adhesion to both plasticware and glass surfaces

  • Synthetic amino acid chain
  • Positively charged having one hydrobromide per unit of Lysine
  • Varying molecular weight: Less viscous lower molecular weight (30,000 d), higher molecular weight (greater than 300,000 d)
  • Uses Poly-L-Lysine of 84000 d, yielding solution viscosity for easy handling while providing sufficient binding sites for cell attachment
  • Product is filtered through a 0.2μm filter before freezing

Specifications

Content And Storage Store at -20°C for up to 4 months from date of receipt. Avoid multiple freeze thaw cycles to maintain product integrity.
Concentration 0.01% w/v (0.1 mg/mL)
Form Liquid
Product Type Poly-L-Lysine
For Use With (Application) Cell Attachment, Neuroscience, Cell Culture
Quantity 100 mL