missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethyleneimine) Solution, 50% (w/v) in Water, MilliporeSigma™ Supelco™

Catalog No. 111020071
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020071 100 mL
111018622 500 mL
111018445 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111020071 Supplier Merck Emd Millipore Supplier No. P3143100ML

Specifications

CAS 9002-98-6
Concentration 50% (w/v) in H 2 O
Synonym Ethyleneimine polymer solution; PEI
Type Analytical Standard
Quantity 100 mL
Grade Analytical Standard