missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene glycol) for GPC 6,000, MilliporeSigma™ Supelco™

Catalog No. 111013733
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013733 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013733 Supplier Merck Emd Millipore Supplier No. 812931G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 25322-68-3
Flash Point 229°C
Physical Form Neat
Linear Formula H(OCH2 CH2)nOH
MDL Number MFCD00081839
Synonym PEG
Type Analytical Standard
Quantity 1 g
Grade GPC