missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene glycol) for GPC 4,000, MilliporeSigma™ Supelco™

Catalog No. 111013729
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013729 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013729 Supplier Merck Emd Millipore Supplier No. 812921G

missing translation for 'validMainframeMsg'

Poly(ethylene glycol) (PEG) is regarded as the gold standard for stealth polymers in polymer based drug delivery. PEGs and their anionic or non-ionic derivatives are widely used in cosmetic products.

Specifications

CAS 25322-68-3
Physical Form Neat
Linear Formula H(OCH2 CH2)nOH
MDL Number MFCD00081839
Synonym PEG
Type Analytical Standard
Quantity 1 g
Grade GPC