missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(ethylene Glycol), Average M.W. 8000
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Supplier: Thermo Scientific Chemicals 418050030
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| white flakes | |
| Average M.W. 8000 | |
| 55.0°C to 60.0°C | |
| H(OCH2CH2)nOH | |
| 15, 7688 | |
| 3 kg | |
| [H]OCCO | |
| 62.07 | |
| 12 to 16mg KOH/g | |
| 300-450 mPa.s (20°C 50% in water) |
| Poly(ethylene glycol) | |
| 25322-68-3 | |
| (C2H4O)n | |
| MFCD01779601 | |
| PEG | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol | |
| 270°C | |
| Plastic drum | |
| ≤0.5% (K.F.) |
Chemical Identifiers
| 25322-68-3 | |
| 62.07 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol |
| (C2H4O)n | |
| MFCD01779601 | |
| PEG | |
| [H]OCCO |
Safety and Handling
EINECSNumber : 500-038-2
RTECSNumber : TQ4100000
TSCA : TSCA
RUO – Research Use Only