missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(ethylene Glycol), Average M.W. 8000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | white flakes |
| Melting Point | 55.0°C to 60.0°C |
| Linear Formula | H(OCH2CH2)nOH |
| Merck Index | 15, 7688 |
| Quantity | 1 kg |
| Flash Point | 270°C |
| Hydroxyl Value | 12 to 16mg KOH/g |
| Packaging | Plastic Bottle |
| Viscosity | 300-450 mPa.s (20°C 50% in water) |
| Water | ≤0.5% (K.F.) |
| Show More |
Safety and Handling
| EINECSNumber | 500-038-2 |
| RTECSNumber | TQ4100000 |
| TSCA | TSCA |
RUO – Research Use Only