missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Poly(acrylic acid) partial sodium salt solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
$369.84 - $369.84
Specifications
| CAS | 9033-79-8 |
|---|---|
| Refractive Index | n20/D 1.418 |
| MDL Number | MFCD00084394 |
| Synonym | Acrylic acid-sodium acrylate copolymer; Sodium acrylate-acrylic acid copolymer |
| Molecular Weight (g/mol) | 5000 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
192031500G
|
Sigma Aldrich
192031500G |
500 g |
Each for $369.84
|
|
|||||
Description
Poly(acrylic acid) partial sodium salt solutionSpecifications
| 9033-79-8 | |
| MFCD00084394 | |
| 5000 |
| n20/D 1.418 | |
| Acrylic acid-sodium acrylate copolymer; Sodium acrylate-acrylic acid copolymer |