missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Poly(acrylic acid) partial sodium salt solution

Catalog No. 192031500G
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
192031500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 192031500G Supplier Sigma Aldrich Supplier No. 192031500G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(acrylic acid) partial sodium salt solution

Specifications

CAS 9033-79-8
Refractive Index n20/D 1.418
MDL Number MFCD00084394
Quantity 500 g
Synonym Acrylic acid-sodium acrylate copolymer; Sodium acrylate-acrylic acid copolymer
Molecular Weight (g/mol) 5000