missing translation for 'onlineSavingsMsg'
Learn More

Poly(acrylic acid), 63 wt% solution in water; approx. M.W. 2,000

Catalog No. p-185690
Change view
Click to view available options
Quantity:
5 g
250 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC184990050 5 g Glass bottle
AC184992500 250 g Glass bottle
AC184995000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC184990050 Supplier Thermo Scientific Chemicals Supplier No. 184990050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9003-01-4 | (C3H4O2)n | 72.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9003-01-4 , 79-10-7 , 7732-18-5
Molecular Formula (C3H4O2)n
Molecular Weight (g/mol) 72.06
MDL Number MFCD00084394
InChI Key NNMHYFLPFNGQFZ-UHFFFAOYSA-M
Synonym acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
PubChem CID 6581
ChEBI CHEBI:18308
SMILES OC(=O)C(-*)C-*

Specifications

Physical Form Liquid
Boiling Point 100.0°C
Density 1.2300g/mL
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula [-CH2CH(CO2H)-]n
Specific Gravity 1.23
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, dioxane
IUPAC Name Poly(acrylic acid)
Percent Purity 60.0 to 65.0% (solids)
Chemical Name or Material Poly(acrylic acid), 63 wt. % Aqueous Solution
Show More Show Less
RTECSNumber AT4680000
TSCA TSCA

RUO – Research Use Only