missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(acrylic acid), 50 wt.% solution in water; approx. M.W. 3,000 to 5,000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| Boiling Point | 100.0°C |
| Linear Formula | [-CH2CH(CO2H)-]n |
| Density | 1.1800g/mL |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, dioxane |
| Quantity | 5 g |
| IUPAC Name | Poly(acrylic acid) |
| Percent Purity | 48 to 52% (solids) |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Show More |
Safety and Handling
| Hazard Category | |
| RTECSNumber | AT4680000 |
| TSCA | TSCA |
RUO – Research Use Only