missing translation for 'onlineSavingsMsg'
Learn More

Poly(3-octylthiophene-2,5-diyl), regiorandom, low metals, Thermo Scientific Chemicals

Catalog No. p-7059524
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4477801 0.25 g
AA4477803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4477801 Supplier Thermo Scientific Chemicals Supplier No. 04477801
Only null left

Conducting polymers

Conducting polymers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104934-51-2
Molecular Formula (C12H18S)n
Molecular Weight (g/mol) 241.44
MDL Number MFCD00241412
InChI Key RXIQSINDOTYIGV-UHFFFAOYSA-N
Synonym poly 3-octylthiophene-2,5-diyl , regiorandom, low metals, poly 3-octylthiophene-2,5-diyl , regioregular, low metals
PubChem CID 121233643
IUPAC Name azane;2,5-dimethyl-3-octylthiophene
SMILES CCCCCCCCC1=C(-*)SC(-*)=C1

Specifications

Physical Form Powder
Solubility Information Soluble in THF, CHCl3
Quantity 0.25 g
Chemical Name or Material Poly (3-octylthiophene-2,5-diyl), Regiorandom, low metals
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only