accessibility menu, dialog, popup

UserName

S∗∗Men

CAS RN 104934-51-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C12H18S)n
InChi Key: —
SMILES: CCCCCCCCC1=C(-*)SC(-*)=C1
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)