missing translation for 'onlineSavingsMsg'
Learn More

Poly(1-vinylpyrrolidone-co-Vinyl Acetate) (Copolymer, 7:3) (50% in Ethanol), TCI America™

Catalog No. P038225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P038225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P038225G Supplier TCI America Supplier No. P038225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25086-89-9
Molecular Formula C10H15NO3
Molecular Weight (g/mol) 197.23
MDL Number MFCD00134018
InChI Key FYUWIEKAVLOHSE-UHFFFAOYSA-N
PubChem CID 270885
IUPAC Name 1-ethenylpyrrolidin-2-one; ethenyl acetate
SMILES CC(=O)OC=C.C=CN1CCCC1=O

Specifications

Color Yellow
Quantity 25 g
UN Number 1170
Physical Form Liquid
Chemical Name or Material Poly(1-vinylpyrrolidone-co-Vinyl Acetate) (Copolymer, 7:3) (50% in Ethanol)
EINECSNumber (6)-0326
RTECSNumber AH3539000
TSCA Yes