Learn More
Poltex Extra Wide Countertop Serological Pipet Holder
Catalog No. 22000188
Encompass
Change view
Click to view available options
Mounting:
Countertop
Catalog No. Mounting
22000188 Countertop
1 options
Catalog No. 22000188 Supplier Poltex Supplier No. XWSRPIPAC
Holder keeps pipets tidy and convenient when used on countertop
  • Shelves are removable for easy cleaning
  • Slanted top allows for extra storage of other items
  • Pocket dimensions:
    • 13.97cm (5.5 in.) wide
    • 27.94cm (11 in.) deep
    • 5.71, 6.98, 8.25, 10.79cm (2.25, 2.75, 3.25 and 4.25 in.) tall
Specifications
For Use With (Equipment) Serological Pipets
Mounting Countertop
Dimensions (L x W x H) 28.39 x 15.25 x 40.64cm (11.18 x 6 x 16 in.)
Product Type Pipet Holder
Shelves 4 Tiers