Learn More
Poltex Serological Pipet Holder With Magnets
Catalog No. 22000186
Encompass
Change view
Click to view available options
Mounting:
Magnetic
Catalog No. Mounting
22000186 Magnetic
1 options
Catalog No. 22000186 Supplier Poltex Supplier No. RGSRPIPACM
Get a Poltex Pipette Holder with $1,500 Purchase
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Strong magnets are interchangeable for mounting on either side
  • Shelves are removable for easy cleaning
  • Magnets are interchangeable for mounting on either side
  • Slanted top allows for extra storage of other items
  • Pocket dimensions:
    • 8.89cm (3.5 in.) wide
    • 27.94cm (11 in.) deep
    • 5.71, 6.98, 8.25, 10.79cm (2.25, 2.75, 3.25 and 4.25 in.) tall
Specifications
For Use With (Equipment) Serological Pipets
Mounting Magnetic
Dimensions (L x W x H) 28.41 x 10.79 x 40.64cm (11.187 x 4.25 x 16 in.)
Product Type Pipet Holder
Shelves 4 Tiers