missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLRMT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552284
Description
Immunogen sequence: ELVYVLFMVK DAGLTPDLLS YAAALQCMGR QDQDAGTIER CLEQMSQEGL KLQALFTAVL LSEEDRATVL KAVHKVKPTF SLPPQLPPPV NTSKLLRDVY AKDGRVSYPK LHLPLKTLQC LFEKQLH Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 65%.
This gene encodes a mitochondrial DNA-directed RNA polymerase. The gene product is responsible for mitochondrial gene expression as well as for providing RNA primers for initiation of replication of the mitochondrial genome. Although this polypeptide has the same function as the three nuclear DNA-directed RNA polymerases, it is more closely related to RNA polymerases of phage and mitochondrial polymerases of lower eukaryotes.Specifications
| POLRMT | |
| Polyclonal | |
| Unconjugated | |
| POLRMT | |
| 1110018N15Rik; 4932416K13; APOLMT; DNA-directed RNA polymerase, mitochondrial; h-mtRPOL; LOW QUALITY PROTEIN: DNA-directed RNA polymerase, mitochondrial; mitochondrial DNA-directed RNA polymerase; MTRNAP; MTRPOL; POLRMT; polymerase (RNA) mitochondrial (DNA directed); RNA polymerase mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5442 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00411 | |
| POLRMT | |
| Recombinant protein corresponding to Human POLRMT. Recombinant protein control fragment (Product #RP-102245). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |