missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ POLR1E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530802
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530802 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530802 Supplier Invitrogen™ Supplier No. PA530802
Only null left

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HepG2, HCT116. Predicted reactivity: Chimpanzee (100%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase I which synthesizes ribosomal RNA precursors (PubMed:24207024). Appears to be involved in the formation of the initiation complex at the promoter by mediating the interaction between Pol I and UBTF/UBF (PubMed:24207024). [UniProt].

Specifications

Antigen POLR1E
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Polr1e
Gene Accession No. Q9GZS1
Gene Alias 53kDa; AU042259; D030019D19Rik; DNA-directed RNA polymerase I subunit E; DNA-directed RNA polymerase I subunit RPA49; FLJ13390; FLJ13970; hypothetical protein LOC511587; Paf53; Polr1e; polymerase (RNA) I associated factor 1; polymerase (RNA) I polypeptide E; polymerase (RNA) I polypeptide E, 53kDa; polymerase (RNA) I subunit E; PRAF1; RGD1565773; RNA polymerase I associated factor (Paf53); RNA polymerase I associated factor 53; RNA polymerase I subunit A49; RNA polymerase I subunit E; RNA polymerase I-associated factor 1; RNA polymerase I-associated factor 53; RP11-405L18.3
Gene Symbols Polr1e
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PRAF1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64425
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less