missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Poloxamer NF (407) (Pluronic F-127)
Description
Poloxamer NF (407) (Pluronic F-127) comes as white prilled or cast solid. Odorless, or has a very mild odor.
Specifications
Specifications
| Chemical Name or Material | Poloxamer NF (407) (Pluronic F-127) |
| Packaging | Jar, 2.8L, White, HDPE, Round, 110-400, TE Pano Neck |
| CAS | 9003-11-6 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 9840 to 14600 (average) |
| Quantity | 1 kg |
| Grade | NF |