missing translation for 'onlineSavingsMsg'
Learn More

Mirus Bio™ pOET6 BacMam Transfer Plasmid

Catalog No. MIR6152
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR6152 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MIR6152 Supplier Mirus Bio™ Supplier No. MIR 6152

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

pOET6 BacMam Transfer Plasmid for high-yield protein production in mammalian cells.

Baculovirus transfer plasmid designed for use with the flashBAC™ Baculovirus Expression System and TransIT™-Insect Transfection Reagent. The CMV promoter enables high-level gene expression in mammalian cells.
  • Simple construction of recombinant virus when used with the flashBAC™ Baculovirus Expression System
  • Can be used with any baculovirus expression system that relies on recombination at the polyhedrin locus
  • Can be propagated in DH5α or other general purpose E. coli strains
Vector Features:
  • Col E1 origin of replication
  • Ampicillin resistance gene for selection in E. coli
  • CMV promoter to drive constitutive foreign gene expression
  • Multiple cloning site containing 11 unique restriction sites for inserting the gene in the correct orientation

Warranty

1 Year

Specifications

Concentration 500 ng/μL, Sterile filtered in TE Buffer
Content And Storage Store at -20°C
Format Kit
For Use With (Application) Protein production in mammalian cells
Product Type Transfer Plasmid
Quantity 20 μL