missing translation for 'onlineSavingsMsg'
Learn More

Mirus Bio™ pOET1C_6xHis Transfer Plasmid

Catalog No. MIR6151
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR6151 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MIR6151 Supplier Mirus Bio™ Supplier No. MIR 6151

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

pOET1C_6xHis Transfer Plasmid for high-yield protein production in insect cells.

Baculovirus transfer plasmid designed for use with the flashBAC™ Baculovirus Expression System and TransIT™-Insect Transfection Reagent. The powerful AcMNPV polyhedrin promoter affords high-level gene expression in insect cells. A C-terminal 6xHis-tag fusion sequence is included for recombinant protein purification with Ni-NTA agarose columns and subsequent removal by thrombin cleavage.
  • Simple construction of recombinant virus when used with the flashBAC™ Baculovirus Expression System
  • Can be used with any baculovirus expression system that relies on recombination in insect cells at the polyhedrin locus
  • Can be propagated in DH5α or other general purpose E. coli strains
Vector Features:
  • Col E1 origin of replication
  • Ampicillin resistance gene for selection in E. coli
  • Multiple cloning site for insertion of the foreign gene in the correct frame and orientation
  • C-terminal 6xHis-Tag™ fusion sequence

Warranty

1 Year

Specifications

Concentration 500 ng/μL, Sterile filtered in TE Buffer
Content And Storage Store at -20°C
Format Kit
For Use With (Application) Protein production in insect cells
Product Type Transfer Plasmid
Quantity 20 μL