missing translation for 'onlineSavingsMsg'
Learn More

PNU 22394 hydrochloride, Tocris Bioscience™

5-HT2C agonist and 5-HT2A/2B partial agonist.

$1469.15 - $1469.15

Chemical Identifiers

CAS 15923-42-9
Molecular Formula C13H17ClN2
Molecular Weight (g/mol) 236.743
InChI Key SAIGGEUWSYESTR-UHFFFAOYSA-N
Synonym pnu 22394 hydrochloride, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indole hydrochloride, 6-methyl-1h,2h,3h,4h,5h-azepino 4,5-b indole hydrochloride, 6-methyl-1,2,3,4,5,6-hexahydroazepino 4,5-b indol-6-ium chloride, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indol, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indole-hydrochloride
PubChem CID 16759162
IUPAC Name 6-methyl-2,3,4,5-tetrahydro-1H-azepino[4,5-b]indole;hydrochloride
SMILES CN1C2=C(CCNCC2)C3=CC=CC=C31.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
2201-10
View Documents
SPEX CertiPrep Group LLC
220110
10 mg
Pack of 10 Each for $1,469.15
Only null left
 
Description

Description

Potent 5-HT2C agonist and partial 5-HT2A/5-HT2B agonist. Nonselective between 5-HT2 receptor subtypes (Ki values are 18, 18 and 66 nM for human recombinant 5-HT2C, 5-HT2A and 5-HT2B receptors respectively). Reduces food intake in rats following subcutaneous administration and displays anorexigenic effects in humans.
Specifications

Chemical Identifiers

15923-42-9
236.743
pnu 22394 hydrochloride, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indole hydrochloride, 6-methyl-1h,2h,3h,4h,5h-azepino 4,5-b indole hydrochloride, 6-methyl-1,2,3,4,5,6-hexahydroazepino 4,5-b indol-6-ium chloride, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indol, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indole-hydrochloride
6-methyl-2,3,4,5-tetrahydro-1H-azepino[4,5-b]indole;hydrochloride
C13H17ClN2
SAIGGEUWSYESTR-UHFFFAOYSA-N
16759162
CN1C2=C(CCNCC2)C3=CC=CC=C31.Cl

Specifications

15923-42-9
10 mg
SAIGGEUWSYESTR-UHFFFAOYSA-N
6-methyl-2,3,4,5-tetrahydro-1H-azepino[4,5-b]indole;hydrochloride
16759162
>99%
C13H17ClN2
pnu 22394 hydrochloride, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indole hydrochloride, 6-methyl-1h,2h,3h,4h,5h-azepino 4,5-b indole hydrochloride, 6-methyl-1,2,3,4,5,6-hexahydroazepino 4,5-b indol-6-ium chloride, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indol, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indole-hydrochloride
CN1C2=C(CCNCC2)C3=CC=CC=C31.Cl
236.743
236.74
PNU 22394 hydrochloride
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Desiccate at +4°C

SDS
missing translation for 'documents'

missing translation for 'documents'