missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PNLIPRP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588845 Supplier Invitrogen™ Supplier No. PA588845
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: LGYPCASYDE FQESKCFPCP AEGCPKMGHY ADQFKGKTSA VEQTFFLNTG ESGNFTSWRY KVSVTLSGKE KVNGYIRIAL YGSNENSKQY EIFKGSLKPD ASHTCAIDVD FNVGKIQKVK FLWNKRGINL SEPKLGASQI TVQSGEDGTE YNFCSSDTVE ENVLQSLYPC; Positive Samples: Rat pancreas.

PNLIPRP2 is a lipase with broad substrate specificity. It can hydrolyze both phospholipids and galactolipids. It acts preferentially on monoglycerides, phospholipids and galactolipids. PNLIPRP2 also contributes to milk fat hydrolysis.

Specifications

Antigen PNLIPRP2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PNLIPRP2
Gene Accession No. P17892, P54318
Gene Alias Cytotoxic T-lymphocyte lipase; galactolipase; pancreatic lipase; pancreatic lipase related protein 2; pancreatic lipase related protein 2 (gene/pseudogene); pancreatic lipase-related protein 2; pancreatic lipase-related protein type 2; pancreatic lipase-related protein type 2 precursor; Plrp2; PL-RP2; PNLIPRP2; Secretory glycoprotein GP-3
Gene Symbols PNLIPRP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 300-469 of human PNLIPRP2 (NP_005387.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117554, 18947
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less