missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PMS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. pipa5102597
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102597 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102597 Supplier Invitrogen™ Supplier No. PA5102597
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PMS1.

This gene encodes a protein belonging to the DNA mismatch repair mutL/hexB family. This protein is thought to be involved in the repair of DNA mismatches, and it can form heterodimers with MLH1, a known DNA mismatch repair protein. Mutations in this gene cause hereditary nonpolyposis colorectal cancer type 3 (HNPCC3) either alone or in combination with mutations in other genes involved in the HNPCC phenotype, which is also known as Lynch syndrome.

Specifications

Antigen PMS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PMS1
Gene Accession No. P54277
Gene Alias DNA mismatch repair protein PMS1; HNPCC3; hPMS1; human homolog of yeast mutL; mismatch repair gene PMSL1; MLH2; PMS1; PMS1 homolog 1, mismatch repair system component; PMS1 postmeiotic segregation increased 1; PMS1 protein homolog 1; PMSL1; postmeiotic segregation increased 1 (S. cerevisiae); rhabdomyosarcoma antigen MU-RMS-40.10B; rhabdomyosarcoma antigen MU-RMS-40.10E
Gene Symbols PMS1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PMS1(Accession P54277), corresponding to amino acid residues N443-I493.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 494322, 5378
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less