missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PluriSIn 1 98.0+%, TCI America™
$259.11 - $771.71
Chemical Identifiers
| CAS | 91396-88-2 |
|---|---|
| Molecular Formula | C12H11N3O |
| Molecular Weight (g/mol) | 213.24 |
| MDL Number | MFCD02344904 |
| InChI Key | HUDWXDLBWRHCKO-UHFFFAOYSA-N |
| Synonym | plurisln 1, n'-phenylisonicotinohydrazide, plurisln 1 nsc 14613, plurisin 1, plurisin #1 nsc 14613, plurisin #1, 4-pyridinecarboxylicacid, 2-phenylhydrazide, 4-pyridinecarboxylic acid 2-phenylhydrazide, plurisln, plurisln1 |
| PubChem CID | 225362 |
| IUPAC Name | N'-phenylpyridine-4-carbohydrazide |
| SMILES | O=C(NNC1=CC=CC=C1)C1=CC=NC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I097520MG
|
TCI America
I097520MG |
20 mg |
Each for $259.11
|
|
|||||
|
I0975100MG
|
TCI America
I0975100MG |
100 mg |
Each for $771.71
|
|
|||||
Chemical Identifiers
| 91396-88-2 | |
| 213.24 | |
| HUDWXDLBWRHCKO-UHFFFAOYSA-N | |
| 225362 | |
| O=C(NNC1=CC=CC=C1)C1=CC=NC=C1 |
| C12H11N3O | |
| MFCD02344904 | |
| plurisln 1, n'-phenylisonicotinohydrazide, plurisln 1 nsc 14613, plurisin 1, plurisin #1 nsc 14613, plurisin #1, 4-pyridinecarboxylicacid, 2-phenylhydrazide, 4-pyridinecarboxylic acid 2-phenylhydrazide, plurisln, plurisln1 | |
| N'-phenylpyridine-4-carbohydrazide |
Specifications
| 91396-88-2 | |
| C12H11N3O | |
| 20 mg | |
| HUDWXDLBWRHCKO-UHFFFAOYSA-N | |
| N'-phenylpyridine-4-carbohydrazide | |
| 225362 | |
| ≥98.0% (HPLC) | |
| PluriSIn 1 |
| 189°C | |
| MFCD02344904 | |
| plurisln 1, n'-phenylisonicotinohydrazide, plurisln 1 nsc 14613, plurisin 1, plurisin #1 nsc 14613, plurisin #1, 4-pyridinecarboxylicacid, 2-phenylhydrazide, 4-pyridinecarboxylic acid 2-phenylhydrazide, plurisln, plurisln1 | |
| O=C(NNC1=CC=CC=C1)C1=CC=NC=C1 | |
| 213.24 | |
| 213.24 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)