missing translation for 'onlineSavingsMsg'
Learn More

Plumbagin, 99+%

Catalog No. AC184040010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC184040010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC184040010 Supplier Thermo Scientific Chemicals Supplier No. 184040010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 481-42-5 | C11H8O3 | 188.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 481-42-5
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.18
MDL Number MFCD00001682
InChI Key VCMMXZQDRFWYSE-UHFFFAOYSA-N
Synonym plumbagin, plumbagine, 5-hydroxy-2-methyl-1,4-naphthoquinone, plumbaein, plumbagone, 2-methyljuglone, 5-hydroxy-2-methyl-1,4-naphthalenedione, 2-methyl-5-hydroxy-1,4-naphthoquinone, 5-hydroxy-2-methyl-1,4 naphthoquinone, 1,4-naphthalenedione, 5-hydroxy-2-methyl
PubChem CID 10205
ChEBI CHEBI:8273
IUPAC Name 5-hydroxy-2-methylnaphthalene-1,4-dione
SMILES CC1=CC(=O)C2=C(C1=O)C=CC=C2O

Specifications

Melting Point 75°C to 78°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 7655
Solubility Information Solubility in water: slightly soluble in hot water. Other solubilities: soluble in alcohol,acetone,chloroform,benzene,,acetic acid
Formula Weight 188.18
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Plumbagin, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-569-6

RUO – Research Use Only