missing translation for 'onlineSavingsMsg'
Learn More

Labcon Plug Style Caps for 12 x 75 mm Culture Tubes

Catalog No. 50101622
Encompass
Change view
Click to view available options
Color:
Assorted
Blue
Green
Purple
Red
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
50101622 Assorted
50101621 Yellow
50101619 Red
50101618 Blue
50101615 Purple
50101616 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 50101622 Supplier Labcon Supplier No. 33028700009

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cap includes flutes to securely grip sides of tubes

  • Fluted Plug Style Cap
  • Plug cap for culture tubes
  • Fits 12 x 75 mm culture tubes
  • Cap includes flutes to securely grip sides of tubes
  • Endotoxin Free (non-pyrogenic)
  • Polyethylene USP Class VI resin
  • Made in USA

Specifications

Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Closure Size 13 mm
Pre-sterilized No
Product Type Tube Plugs
Autoclavable Not Autoclavable
Material Polyethylene
Color Assorted
For Use With (Application) Centrifugation; For Cell Culture and General Laboratory Use
Clean Claims ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
Length (Metric) 75 mm
Sterility Non-sterile
For Use With (Equipment) Tubes
Disposable Single-use
Show More Show Less