missing translation for 'onlineSavingsMsg'
Learn More

Accuform Signs Plug Lockouts

Catalog No. 19821771
Change view
Click to view available options
For Use With (Equipment):
Cord size up to 0.5 in. dia., plug size up to 3.25 x 1.75 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19821771 Cord size up to 0.5 in. dia., plug size up to 3.25 x 1.75 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19821771 Supplier Accuform Signs Supplier No. KDD237
Only null left

Hinge style lockouts seal off plug to prevent equipment from being plugged in

Specifications

Product Type Hinged Plug Lockout
Color Red
Material Plastic
For Use With (Equipment) Cord size up to 0.5 in. dia., plug size up to 3.25 x 1.75 in.