missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLIN4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150273
Description
PLIN4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Chemical Identifiers
| PLIN4 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q96Q06 | |
| 100524667, 729359 | |
| Recombinant Perilipin 4 (PLIN4) (Leu349-Gly656). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Pig | |
| PLIN4 | |
| adipocyte protein S3-12; KIAA1881; perilipin 4; perilipin-4; PLIN4; S3-12 | |
| PLIN4 | |
| Primary | |
| Protein A |
Specifications
| PLIN4 | |
| Polyclonal | |
| Unconjugated | |
| PLIN4 | |
| adipocyte protein S3-12; KIAA1881; perilipin 4; perilipin-4; PLIN4; S3-12 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100524667, 729359 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q96Q06 | |
| PLIN4 | |
| Recombinant Perilipin 4 (PLIN4) (Leu349-Gly656). | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody |