missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLEKHG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553684
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553684 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553684 Supplier Invitrogen™ Supplier No. PA553684
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: ATRRELFSGS NPGKLGEPPS GGKAGPEEDE EGVSFTDFQP QDVTQHQGFP DELAFRSCSE IRSAWQALEQ GQLARPGFPE PLLILEDSDL GGDSGSGKAG APSSERTASR VRELARLYSE RI Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 77%.

PLEKHG2 may be a transforming oncogene with exchange activity for CDC42. PLEKHG2 may be a guanine-nucleotide exchange factor (GEF) for RAC1 and CDC42.PLEKHG2 activated by the binding to subunits beta and gamma of the heterotrimeric guanine nucleotide-binding protein (G protein).

Specifications

Antigen PLEKHG2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLEKHG2
Gene Accession No. Q9H7P9
Gene Alias AI194308; ARHGEF42; Clg; common site lymphoma/leukemia GEF; Common site lymphoma/leukemia guanine nucleotide exchange factor; common-site lymphoma/leukemia GEF; common-site lymphoma/leukemia guanine nucleotide exchange factor; Cslg; CTB-60E11.4; DKFZp667J2325; FLJ00018; FLJ22458; FLJ38638; LDAMD; LOW QUALITY PROTEIN: pleckstrin homology domain-containing family G member 2; PH domain-containing family G member 2; pleckstrin homology and RhoGEF domain containing G2; pleckstrin homology domain containing, family G (with RhoGef domain) member 2; pleckstrin homology domain-containing family G member 2; Plekhg2; RGD1310112
Gene Symbols PLEKHG2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLEKHG2. Recombinant protein control fragment (Product #RP-90704).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64857
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less