missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pleiotrophin Monoclonal Antibody (K06279_4G7), Invitrogen™
Mouse Monoclonal Antibody
$1421.98 - $2134.69
Specifications
| Antigen | Pleiotrophin |
|---|---|
| Clone | K06279_4G7 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Pleiotrophin Monoclonal antibody specifically detects Pleiotrophin in Human, Mouse samples. It is validated for Western BlotSpecifications
| Pleiotrophin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| HARP; HBBM; HBBN; HB-GAM; HBGF8; HBGF-8; HBNF; HBNF1; HBNF-1; heparin affin regulatory peptide; heparin affin regulatory protein; heparin binding factor; heparin-binding brain mitogen; Heparin-binding growth factor 8; heparin-binding growth-associated molecule; heparin-binding neurite outgrowth promoting factor; Heparin-binding neurite outgrowth-promoting factor; Heparin-binding neurite outgrowth-promoting factor 1; heparin-binding neurite promoting factor; heparin-binding neurotorphic factor; heparin-binding neutrophic factor; NEGF1; OSF; Osf1; OSF-1; osteoblastic cell factor; Osteoblast-specific factor 1; pleiotrophin; pleiotrophin (heparin binding growth factor 8, neurite growth-promoting factor 1); Pleiotrophin (Heparine binding factor, Hbnf, in the mouse); PTN | |
| Recombinant human PTN (proprietary range information). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| PTN |
| K06279_4G7 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| P21246, P63089 | |
| 19242, 5764 | |
| IgG1 | |
| Protein A | |
| Antibody |