missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLCH2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149857
Description
PLCH2 Polyclonal Antibody for Western Blot, ICC/IF, IHC
PLCH2 is a member of the PLC-eta family of the phosphoinositide-specific phospholipase C (PLC) superfamily of enzymes that cleave PtdIns(4,5) P2 to generate second messengers inositol 1,4,5-trisphosphate and diacylglycerol (Zhou et al., 2005 [PubMed 16107206]).Specifications
| PLCH2 | |
| Polyclonal | |
| Unconjugated | |
| PLCH2 | |
| 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase eta-2; A930027K05Rik; KIAA0450; Phosphoinositide phospholipase C-eta-2; phosphoinositide phospholipase C-like 4; phospholipase C eta 2; phospholipase C, eta 2; phospholipase C-eta2; Phospholipase C-eta-2; phospholipase C-like 4; phospholipase C-like protein 4; PLCeta2; Plc-eta2; PLCH2; PLCL4; PLC-L4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 269615, 313756, 9651 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| A2AP18, O75038 | |
| PLCH2 | |
| Recombinant Phospholipase C Eta 2 (PLCh2) (Met1-His257). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |