missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLCE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150003
Description
PLCE1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a phospholipase enzyme that catalyzes the hydrolysis of phosphatidylinositol-4,5-bisphosphate to generate two second messengers: inositol 1,4,5-triphosphate (IP3) and diacylglycerol (DAG). These second messengers subsequently regulate various processes affecting cell growth, differentiation, and gene expression. This enzyme is regulated by small monomeric GTPases of the Ras and Rho families and by heterotrimeric G proteins. In addition to its phospholipase C catalytic activity, this enzyme has an N-terminal domain with guanine nucleotide exchange (GEF) activity. Mutations in this gene cause early-onset nephrotic syndrome; characterized by proteinuria, edema, and diffuse mesangial sclerosis or focal and segmental glomerulosclerosis. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Chemical Identifiers
| PLCE1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8K4S1, Q99P84, Q9P212 | |
| 114633, 51196, 74055 | |
| Recombinant Phospholipase C Epsilon 1 (PLCe1) (Met1-Cys250). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| PLCE1 | |
| 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase epsilon-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase epsilon-1; 4933403A21Rik; KIAA1516; mKIAA1516; NPHS3; pancreas-enriched phospholipase C; phosphoinositide phospholipase C; phosphoinositide phospholipase C-epsilon-1; phosphoinositide-specific phospholipase C epsilon-1; phospholipase C epsilon 1; phospholipase C, epsilon 1; phospholipase C-epsilon-1; PLCE; PLCE1; PLCepsilon; PLC-epsilon-1; PPLC | |
| PLCE1 | |
| Primary | |
| Protein A |
Specifications
| PLCE1 | |
| Polyclonal | |
| Unconjugated | |
| PLCE1 | |
| 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase epsilon-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase epsilon-1; 4933403A21Rik; KIAA1516; mKIAA1516; NPHS3; pancreas-enriched phospholipase C; phosphoinositide phospholipase C; phosphoinositide phospholipase C-epsilon-1; phosphoinositide-specific phospholipase C epsilon-1; phospholipase C epsilon 1; phospholipase C, epsilon 1; phospholipase C-epsilon-1; PLCE; PLCE1; PLCepsilon; PLC-epsilon-1; PPLC | |
| Rabbit | |
| Protein A | |
| RUO | |
| 114633, 51196, 74055 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8K4S1, Q99P84, Q9P212 | |
| PLCE1 | |
| Recombinant Phospholipase C Epsilon 1 (PLCe1) (Met1-Cys250). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |