missing translation for 'onlineSavingsMsg'
Learn More

Platinum(IV) oxide monohydrate, 99.8%, (trace metal basis)

CAS: 12137-21-2 | H2O3Pt | 245.10 g/mol

$147.66 - $498.97

Chemical Identifiers

CAS 12137-21-2
Molecular Formula H2O3Pt
Molecular Weight (g/mol) 245.10
MDL Number MFCD00066964
InChI Key SPGAMGILENUIOF-UHFFFAOYSA-N
Synonym dioxoplatinum hydrate, platinum iv oxide monohydrate, platinum dioxide hydrate, platinum iv oxide hydrate, platinoxydhydrat, acmc-1bujp, platinum 1v oxide hydrate, pto2 h2o
PubChem CID 11139377
SMILES O.O=[Pt]=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC211932500
SDS View Documents
Thermo Scientific Chemicals
211932500
250 mg Glass bottle
Each for $147.66
Only null left
 
AC211930010
SDS View Documents
Thermo Scientific Chemicals
211930010
1 g Glass bottle
Each for $498.97
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
Specifications

Chemical Identifiers

12137-21-2
245.10
SPGAMGILENUIOF-UHFFFAOYSA-N
11139377
H2O3Pt
MFCD00066964
dioxoplatinum hydrate, platinum iv oxide monohydrate, platinum dioxide hydrate, platinum iv oxide hydrate, platinoxydhydrat, acmc-1bujp, platinum 1v oxide hydrate, pto2 h2o
O.O=[Pt]=O

Specifications

12137-21-2
Powder
99.8%
PtO2·H2O
01,890
dioxoplatinum hydrate, platinum iv oxide monohydrate, platinum dioxide hydrate, platinum iv oxide hydrate, platinoxydhydrat, acmc-1bujp, platinum 1v oxide hydrate, pto2 h2o
SPGAMGILENUIOF-UHFFFAOYSA-N
dioxoplatinum;hydrate
11139377
99.80%
Glass bottle
Brown to Black
250 mg
H2O3Pt
MFCD00066964
10, 7398
Solubility in water: insoluble in water. Other solubilities: soluble in conc.acids and in dil.sol. of koh
O.O=[Pt]=O
245.10
245.1
Trace Metal Basis
Platinum(IV) oxide monohydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause fire or explosion; strong oxidizer.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only